Work caps, PP
Catalog # HOIT115799
Supplier: HUBEI ORIENT INTL TRADING
Specifications
- Applications des tenues:Alimentaire
- Genre:
- Désignation:Work caps, 40 g/m²
- Environmentally Preferable:
- Cdt:1.000
Specifications
About this item
Caps made of nonwoven material.
- Lightweight
- Breathable.
Keep off hair or dandruff from the work area effectively.
Weight: 90 g/m² (Peak), 35 g/m² (Caps).
Packaging: Package of 100/bag, 10 bags/carton.