Laboratory coats
Laboratory coats
Katalognr.
Laboratory coats
Katalognr.
Anbieter:  
Laboratory coats
Laboratory coats
Product Details & Documents
Ideal for non critical application.
  • Snap style
  • Made of polypropylene
  • Two pocket lab coat
The collar of shirt comes in Shirt, Korean, Knitted type. These coats are available in white/green/blue colour with size from S to 5XL.
Weight: 25 to 55 g/m²