ContactUsLinkComponent
Pharmaceutical secondary standard, Zidovudine, certified reference material, Supelco®
  PHR1292-1G
 :  Merck
 :  
undefined
Pharmaceutical secondary standard, Zidovudine, certified reference material, Supelco®
  PHR1292-1G
 :  Merck
 :  
  • Concentration:
    Neat
  • Cdt:
    1 g
  • Normes:
    Pharmaceutical secondary standard (CRM)
  • N ° CAS.:
    30516-87-1
  • Désignation:
    Zidovudine
  • Désignation:
    Zidovudine
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
 : This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.