Pharmaceutical secondary standard, Zidovudine, certified reference material, Supelco®
PHR1292-1G
: Merck
:
- Concentration:Neat
- Cdt:1 g
- Normes:Pharmaceutical secondary standard (CRM)
- N ° CAS.:30516-87-1
- Désignation:Zidovudine
- Désignation:Zidovudine
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.