CytoRAL fixative for cytological smears, RAL
Catalog # 720-1153
Supplier: RAL
CAS Number:
Specifications
- Cdt:1
- Désignation:Fixing spray 150 ml
Specifications
About this item
CytoRAL is used for cervical smears, urine, sputa and broncho-alveolar fluids.
It lays a protective film on the smears, keeping a good hydration before staining and without any alteration of the original cells aspect. With a large dispersion ratio the spray covers the entire slide area.