Cervical brushes, Cytotak™ Transwab®, labelled
720-0136
- Handle material:Plastique
- Désignation:Cytotak, tube étiqueté
- Seq:01
- Longueur totale:150 mm
- Cdt:100
Nylon-bristle cytology sampling brushes for cervical smears.
- Sterile, packed in labelled tube
- Plastic shaft is non toxic and safe to use
- Tapered spiral configuration assists access to narrow opening
- Gentle rotation creates a brushing action against skin surface for enhanced recovery of cellular material
Shaft/head: Plastic/nylon
: Individually packed.