ContactUsLinkComponent
Touch-free soap dispenser, MicroDispenser™
Touch-free soap dispenser, MicroDispenser™
Référence MICRICDISP-4
Fournisseur:  MICRONOVA
undefined
Touch-free soap dispenser, MicroDispenser™
Référence MICRICDISP-4
Fournisseur:  MICRONOVA
Spécifications
  • Capacité:
    500 ml
  • Désignation:
    Touch-free dispenser for Micronova 500 ml pump bottles
  • Cdt:
    1
Spécifications
À propos de cet article
Touch-free soap dispensing unit effectively reduces cross-contamination and waste in the cleanroom and gowning areas.
  • Reduces cross-contamination and waste
  • Activated by an electronic eye
  • Delivers a consistent, premeasured dose of soap, lotion, or sanitiser into the palm
  • Gravity fed with no tubes or pumps
  • No-drip unit may be wall-mounted
Easy delivery system allows dispenser to be use for different soaps or lotions by changing spout. Can be used with 500 ml pump bottles.
Certifications: Complies with OSHA, CDC, and ADA requirements.
Livraison: Operates on four size AA batteries which are included.