ASTM® D5442 quantitative linearity standard, C12 - C50, analytical standard, Supelco®
Catalog # 502243.
Supplier: Merck
CAS Number:
Specifications
- Concentration:0.01 % (w/w) each component in cyclohexane
- Cdt:1
- Désignation:ASTM(R) D5442 Quantitative Linearity Standard, C12 - C50