Alkane standard mixture for performance tests of GC-systems, C10 - C40 (all even), analytical standard, Supelco®
Catalog # 68281-2ML-F
Supplier: Merck
CAS Number:
Specifications
- Concentration:50 mg/l each
- Cdt:2 mL
- Désignation:Alkane standard mixture for performance tests of GC-systems, C10 - C40 (all even)
- M-Clarity quality level:MQ100