Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Étalons
Étalons de turbidité
Turbidity - Whole Volume - WP, Proficiency Testing Material, Supelco®
Turbidity - Whole Volume - WP, Proficiency Testing Material, Supelco®
Turbidity - Whole Volume - WP, Proficiency Testing Material, Supelco®
Catalog # PE3081-500ML
Supplier:
Merck
CAS Number:
Expand image
Turbidity - Whole Volume - WP, Proficiency Testing Material, Supelco®
Catalog # PE3081-500ML
Supplier:
Merck
CAS Number:
Specifications
Cdt:
500 mL
Désignation:
Turbidity - Whole Volume - WP
Type:
Proficiency Testing Material
Specifications
About this item
Product Details
Product Details