EPA SS TCL Volatiles mix 1, certified reference material, Supelco®
Catalog # 4S8949
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 mug/mL each component in methanol: water (9:1)
- Cdt:1
- Désignation:EPA SS TCL Volatiles Mix 1
- Type:Certified reference material