Poly(dimethylsiloxane) analytical standard, for GPC
Catalog # 81153-100MG
Supplier: Merck
CAS Number:
Specifications
- Cdt:100 mg
- Désignation:311 Mw, Mw/Mn ~1,00
- M-Clarity quality level:MQ100
Specifications
About this item
Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.
Certifications: Exact values can be taken from the accompanying certificate of analysis
Caution: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.