Disinfectant cleaner, Oxivir Excel
DIVY100941436
: Diversey
- Désignation:Oxivir Excel
- Environmentally Preferable:Safer Human and Environmental Health
- Emballage:2×5 l canisters
- Cdt:2
Disinfectant cleaner with AHP technology. Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
- For surfaces and non invasive medical devices
- Bactericidal, virucidal, fungicidal and yeasticidal activity
- VAH listed, medical device
- Environmentally friendly
- Excellent user safety profile reduces worker risk and absence
Oxivir Excel is a highly concentrated disinfectant cleaner with a broad spectrum of action and short exposure times. Oxivir excel cleans and disinfects in one operation and can be used on all common surfaces, as well as some textiles. The broad spectrum of effects ensures the highest possible patient safety. Extensively tested to European standards, Oxivir Excel provides effective protection against a variety of pathogens, including poliovirus, adenovirus, and murine norovirus. Also effective against enveloped viruses such as HIV, HBV, HCV, influenza, vaccinia virus and the coronavirus.
: Efficacy proven by compliance with: EN 13727, EN 13624, EN 14476, EN 1276 and EN 1650.