Foot pedal for Dino-Lite microscopes, SW-F1
Catalog # 630-1821
Supplier: Dino-Lite / IDCP B.V.
Specifications
- Désignation:Foot pedal, enables users to take a picture with their foot
- Seq:956
- Cdt:1
Specifications
About this item
The SW-F1 allows users to take a picture with a simple step from their foot using the Dino-Lite or Dino-Eye digital microscopy solutions. The foot pedal is only compatible with DinoCapture 2.0 for the PC and DinoXcope for Macintosh computers. No additional software is required, simply plug in the device into your USB port and it is ready to use.
Compatible with all USB models.