Poly(ethylene glycol) analytical standards, for GPC, Supelco
Référence 81201-500MG
Fournisseur: Merck
Numéro CAS:
Spécifications
- Cdt:500 mg
- Désignation:∼96500 Mn, Mw/Mn ~1,12
- M-Clarity quality level:MQ100
Spécifications
À propos de cet article
Poly(ethylene glycol) is the most widely used non-ionic, hydrophilic polymer, which can find wide applications in biomedical industry such as in drug delivery systems.
Certifications: Exact values can be taken from the accompanying certificate of analysis