Chromium VI -WP, certified reference material, Supelco®
Catalog # QC1088-20ML
Supplier: Merck
CAS Number:
Specifications
- Cdt:20 mL
- Élement:Chromium VI
- Symbole:Cr
Specifications
About this item
ICP and AAS suitable.
Values of analytes vary lot to lot.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.