ESD safety shoes, lace-up, uvex 2 xenova®, 9564
UVEX9564.3/46
: UVEX
- Pointure:46
- Couleur:vert/noir
- Cdt:1 PAIR
Very lightweight water repellent uvex 2 xenova® shoes are made from full-grain smooth leather upper, soft padded collar and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances and suitable for people allergic to chrome.
- Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
- Anti-twist rear cap provides more stability and kink protection
- Climazone technology for improved comfort
- Anti-static comfort insole with moisture control system; orthopaedic insoles possible
- Heat-resistant loop lacing system
- DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Shock absorbing heel and forefoot
- Metal-free and penetration-resistant uvex xenova® midsole
- uvex xenova® composite toe cap (200 J)
- Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
Colour: Green/black
: EN 61340 (ESD), EN ISO 20345:2011; S3, SRC