Formaldehyde in Water, (CRM), Supelco®
Katalognr. SUPLQC1380-20ML
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Type de conditionnement:Ampoule
- Cdt:20 mL
- Désignation:Formaldehyde in Water
- Type:Certified reference material
Technische Daten
Über diesen Artikel
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.