Air flow comboKit 2 with Bluetooth®, Testo 440
Catalog # TEST0563.4407
Supplier: Testo
Specifications
- Interface:Bluetooth
- Désignation:Air velocity and IAQ measuring instrument, testo 440 air flow ComboKit 2 with Bluetooth, with internal memory and data export function; 100 mm vane probe with Bluetooth, incl. temperature sensor, 16 mm vane probe with telescope (0,85 m), fixed cable (1,8 m), Combi case for testo 440 and multiple probes
- Seq:77
- Cdt:1
Specifications
About this item
Airflow Combokit 2 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
- Long-term monitoring of indoor air quality
- Combine the vane probe with Bluetooth with the telescope for ceiling outlets
- Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Convenient for wireless probe, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outlet measurements.
Delivery: Supplied with testo 440 air velocity and IAQ measuring instrument, the vane probe (Ø 100 mm) with Bluetooth® including temperature sensor and the vane probe (Ø 16 mm).