silv-Air 7312 Hygiene Masks
N° de catalogue UVEX8707.312
Fournisseur: UVEX
Spécifications
- Type:FFP3 R D preformed mask with valve
- Couleur:blanc
- Cdt:5
Spécifications
À propos de cet article
This FFP3 preformed mask with an exhalation valve, featuring an innovative design and an optimised shape.
- Four-point headband attachment for a comfortable, individual and secure fit
- Extra-wide fabric headband
- Ultrafine porous sealing lip
- Nose clip integrated into the material for comfortable and secure positioning of the mask
The three- dimensional design enables a perfect fit for the many different face shapes which exist.