Screw cap with septum for sample tubes
SIMPT504AQX
- Désignation:Septum screw cap for sample tubes
- Matière:PP
- Cdt:250
This aqua colour screw cap incorporates a pierceable septum made of chemically resistant PTFE on the outside and silicone on the inside, both components being stable over a broad range of temperatures.
- Acts as a silicone O-ring for better sample protection
- Especially made to fit and be used with all Simport® sample tubes
The cap is pierceable with pipette tips as well as with syringe needles.