Cryoembedding media for microscopy, M-Freeze™
Catalog # 1.03693.0100
Supplier: Merck
CAS Number:
Specifications
- Cdt:100 mL
- Désignation:Cryoembedding media for microscopy, M-Freeze™
Specifications
About this item
Cryoembedding media for microscopy M-Freeze™ is used in the routine for preparing specimens of unfixed tissue (native material) of human origin for frozen-section analysis. It is also used for preparing cryo sections for enzyme and immunohistology methods to avoid fixation artefacts.
Delivery information: Supplied ready to use.