Disinfectant detergent, clearKlens deogen
Catalog # DIVY7522374
Supplier: Diversey
Specifications
- Désignation:ClearKlens Deogen VH47
- Specific gravity:1,170 – 1,210 g/ml
- Cdt:20 L
- Valeur pH:12,2
Specifications
About this item
Chlorinated alkaline detergent and disinfectant for soft/medium hard water.
- Low foaming under conditions of high turbulence
- High active chlorine level effectively removes soils, stains and off-flavours
- Broad spectrum disinfectant action
- Free-rinsing, minimises water usage
Used for CIP and spray washing in pharmaceutical, cosmetic, and other industries.
Caution: Utilisez les biocides en toute sécurité. Toujours lire l'étiquette et les informations du produit avant utilisation.