Preservative, grotan® OK
N° de catalogue SCUEGROTANOK
Fournisseur: SCHÜLKE & MAYR GMBH
Spécifications
- Désignation:grotan® OK
- Cdt:10 L
Spécifications
À propos de cet article
grotan® OK is suitable for use in water-mixed metalworking fluids, metalworking fluid concentrates and other technical emulsions.
- Bactericidal effect
- Broad, balanced spectrum of effect (incl. sulphate-reducing bacteria)
- Immediate effect with improved long term efficacy
- Demonstrable effective against Mycobacteria
- Good anticorrosion properties
Contains no nitrate, nitrosing agents or organically bound chlorine (has no effect on the AOX value). Odour improved methylenebisoxazolidine.