Jar, PP, with screw cap, SP Bel-Art
Catalog # BELAH179100000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø ext.:58 mm
- Capacité:118 ml
- Hauteur:66,5 mm
- Environmentally Preferable:
- Cdt:12
Specifications
About this item
White PP jar with screw cap in an assortment of sizes and is used for storage of liquid or dry stock, or for use with samples.
Screw cap with PE foam lining and are not autoclavable.