Human recombinant IL-2 (fromE. coli)
Supplier: PeproTech, Inc.
Certificates
About this item
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation.
IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response.
IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant human IL-2 is a 15,5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
Specifications
- Conjugaison:Unconjugated
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Activité biologique:≥1×107 units/mg
- Réactivité croisée:Bacteria, chicken, cow, cow and mouse, dog, human, human and mouse, human and virus, leech, monkey, mouse, rat
- Synonyme de protéine:Aldesleukin|T-cell Growth Factor (TCGF)
- Nom de la protéine/du peptide:IL-2
- Purity:>98%
- Molecular weight:15.5 kDa
- Sequence:MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFAQSII STLT
- Niveau d'endotoxine:Low
- Purification:Gel and HPLC analyses
Specifications
Frequently Bought Together