Aprons, zetDress®
N° de catalogue ZVGA80160
Fournisseur: Zellstoff-Vertriebs-GmbH & Co. KG
Spécifications
- Anti-statique:+
- Applications des tenues:À usage universel,Laboratoire,Industriel,Alimentaire
- Couleur:blanc
- Cdt:100
- l×L:750×1250 mm
Spécifications
À propos de cet article
Aprons, made from PE foil, are suitable for sensitive work areas, such an the food processing industry, the sustainable maintenance of a hygienic environment, is indispensable.
- Robust and disposable
- Hammered foil
- Individually folded for easy removal
Protects from biological contaminants, radioactive particles and liquid chemicals, also anti-static, spray-proof and particle-tight.