Precision laboratory thermometer
Catalog # SCNL4730732
Supplier: Ludwig Schneider GmbH & CO. KG
Specifications
- Ø:6 mm
- Profondeur d'immersion:Total
- Désignation:Enclosed type, with colourless prismatic capillary form
- Longueur:175 mm
- Environmentally Preferable:
- Division:0,2 °C
- Remplissage:Blue
- Cdt:1
- Plage de mesure:0...+50 °C
Specifications
About this item
These total immersion thermometers are suitable for laboratory purposes.