HEPES (Acide 4-(2-hydroxyéthyl)pipérazine-1-yléthanesulfonique)
Supplier: EUROCLONE
Certificates
About this item
Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
CAS Number:7365-45-9
:
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1.448 g/cm³ (20 °C)
:238.31 g/mol
Storage Temperature:Température ambiante
EINECS:230-907-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together