IHC tissue primer
IHC tissue primer
IHC tissue primer, 100 mL
N° de catalogue ENZOADI9502340100
Fournisseur:   Enzo Life Sciences
IHC tissue primer
N° de catalogue ENZOADI9502340100
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
IHC tissue primer
IHC tissue primer, 100 mL
Numéro CAS:  
Prix
SFr. 284.00
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents
Immunohistochemistry tissue primer is an enzyme blocking reagent.
  • TRIS-based alkaline solution with 30% H₂O₂
Designed for use in horse radish peroxidase (HRP) and alkaline phosphatase (AP) based immunohistochemical staining procedures on paraffin tissue sections, frozen tissue sections and cell smears to suppress non specific staining due to endogenous enzyme activity.
Spécifications
Spécifications
  • Cdt:
    100 mL
  • Désignation:
    IHC tissue primer