Yeast extract, molecular biology grade
A3732.0500
:
- Type de conditionnement:Flacon verre pour solides
- Cdt:500 g
- Désignation:Yeast extract, molecular biology grade
Water-soluble part of autolysed yeast. Rich of ammonia and vitamins, especially of water-soluble vitamin B-complexes; suitable for addition to media.