Dichlorométhane-D2
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2
- CAS Number:1665-00-5
- Formula:CD₂Cl₂
- MDL Number:MFCD00000882
- Molecular Weight:86.92 g/mol
- Boiling Pt:40 °C (1013 hPa)
- Melting Pt:–97 °C
- Density:1,36 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:216-776-0
- UN:1593
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2
- CAS Number:1665-00-5
- Formula:CD₂Cl₂
- MDL Number:MFCD00000882
- Molecular Weight:86.92 g/mol
- Boiling Pt:40 °C (1013 hPa)
- Melting Pt:–97 °C
- Density:1,36 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:216-776-0
- UN:1593
- ADR:6.1,III
Product Family Options
Product Information