Trousers, PP, Hopen®
HOPE40.170/04
: Medicom
- Taille de confection:Taille unique
- Applications des tenues:À usage universel,Médical,Laboratoire,Industriel
- Genre:Unisex
- Poches:sans poches
- Couleur:blanc
- Cdt:50
- Longueur de manches ou de jambes:Long
- Poids:45 g/m²
These disposable PP trousers are ideal for use in combination with PP lab coats.
- Breathable and lightweight fabric
- Easy to put on
- Elasticated waist and ankles
- One size fits all (size U*)
Weight: 45 g/m²
: Cat. I
: Individually packed.