Racks for Gilson Pipetman pipette tips
Catalog # GREI973270_50
Supplier: Greiner Bio-One
Specifications
- Désignation:Empty PP rack for pipette tips (10 - 200 µl), suitable for Gilson®
- Cdt:50
Specifications
About this item
Empty racks for 10 - 200 µl pipette tips that are ideally suited for Gilson Pipetman, Biohit, Brand, Eppendorf, Gilson and Socorex pipettes.