ContactUsLinkComponent
CinnGEL 2-methylester
CinnGEL 2-methylester
Catalog # ENZOBMLPR1150001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
CinnGEL 2-methylester
Catalog # ENZOBMLPR1150001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Cdt:
    1 mg
  • Seq:
    02
  • Nom de la protéine/du peptide:
    CinnGEL 2-methylester
Specifications
About this item
PTP1B inhibitor
Novel peptoid inhibitor of PTP1B (IC50=1,3 µM). Methyl ester (for cell permeability) is hydrolyzed to active inhibitor by intracellular esterases.