Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant IL-1alpha
Human recombinant IL-1alpha
Catalog # ENZOENZPRT1870010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant IL-1alpha
Catalog # ENZOENZPRT1870010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Synonyme de protéine:
    Hematopoietin-1|Endogenous pyrogen|IL-1F1|LAF|Leukocyte endogenous mediator|LEM|Interleukin-1alpha|MCF|Lymphocyte-activating factor |Mononuclear cell factor
  • Nom de la protéine/du peptide:
    IL-1alpha
  • Purity:
    >98% (SDS and HPLC)
  • Molecular weight:
    ~17,9 kDa
  • Formulation:
    Lyophilized from 1 mM TRIS, pH 8,0.
  • Purification:
    Sterile filtered through a 0,2 micron filter.

Specifications

About this item

Produced in E. coli.