ContactUsLinkComponent
Human recombinant IL-7
Catalog # ENZOENZPRT1370010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant IL-7
Catalog # ENZOENZPRT1370010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Synonyme de protéine:
    Interleukin-7
  • Nom de la protéine/du peptide:
    IL-7
  • Purity:
    >98% (SDS-PAGE gel & HPLC)
  • Molecular weight:
    ~17.4 kDa
  • Formulation:
    Lyophilized from 10 mm acetic acid
Specifications
About this item
Produced in E.coli.