Monkey recombinant NGAL
ENZOENZPRT1181000
:
- Cdt:1 mL
- Type de protéine/peptide:Recombinant
- Species:Monkey
- Seq:02
- Synonyme de protéine:Neutrophil gelatinase-associated lipocalin|Siderocalin|Lipocalin 2|Oncogene 24p3
- Nom de la protéine/du peptide:NGAL
- Concentration:1000 pg/ml
- Formulation:Liquid. In 0,01 M phosphate buffer, pH 7,4, containing 0,14 M NaCl, 0,5% BSA, 0,5% Tween 20, 0,0016% Phenol red, and 0,1% Proclin 950
Produced in E. coli.