Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant PKCζ,GST tag
Human recombinant PKCζ,GST tag
Catalog # ENZOBMLSE4130005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant PKCζ,GST tag
Catalog # ENZOBMLSE4130005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Cdt:
    5 µG
  • Type de protéine/peptide:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    GST Tag
  • Nom de la protéine/du peptide:
    PKCζ

Specifications

About this item

Produced in insect cells.