ContactUsLinkComponent
Toxin A
  ENZOALX630100C100
 :  
Toxin A
  ENZOALX630100C100
 :  
  • Cdt:
    100 µG
  • Seq:
    02
  • Nom de la protéine/du peptide:
    Toxin A
Cytotoxic and enterotoxic
Isolated from Clostridium difficile.
Has potent cytotoxic and enterotoxic properties. Translocates to the cytosol of target cells and glucosylates small GTP-binding proteins such as Rho, Rac, and Cdc42. Induces actin condensation, cell death and opening of the tight junctions. Triggers release of pro-inflammatory cytokines from intestinal epithelial cells probably via activation of MAP kinases.