Chemiluminescent labelling kit
Catalog # ENZOADI907001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:Chemiluminescent labelling kit
- Seq:02
Specifications
About this item
A complete kit for the labeling of antibodies, proteins, peptides, and nucleic acids with Acridinium NHS Ester.
Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and peptides containing primary amine groups. These chemiluminescent molecules are frequently for use as detection reagents in immunoassays as a safe alternative to radioactive tracers. The Chemiluminescent labeling kit provides the necessary components for quick, efficient, and easy acridinium labeling.
Ordering information: Contains Acridinium NHS Ester, Dry DMF, Bicarbonate labeling buffer, 10% Lysine, Gel filtration column, Column buffer concentrate, Set of Trigger solutions.