AMPIGENE® Taq DNA polymerase
Catalog # ENZOENZPRT1000500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:AMPIGENE® Taq DNA polymerase
- Seq:02
- Systèmes:500
Specifications
About this item
Optimised standard PCR for enhanced speed, yield, and specificity.
AMPIGENE® Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. It can be used for routine PCR applications including genotyping, screening, and library construction.
Ordering information: Contains AMPIGENE® Taq DNA polymerase (5u/µl)and 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).