Glutathione peroxidase activity kit
N° de catalogue ENZOADI900158
Fournisseur: ENZO LIFE SCIENCES
Numéro CAS:
Spécifications
- Cdt:1 KIT
- Désignation:Glutathione peroxidase activity kit
- Seq:02
- Taille:96 wells
- Tests par set:5×96 wells
Spécifications
À propos de cet article
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes.
For the measurement of glutathione peroxidase in cell lysates, plasma, and tissue from any species.
Informations de commande: Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG