Plate sealing film, AeraSeal
AC1201-02
- Stérile:+
- Désignation:AeraSeal film sheets
- Épaisseur:114 µm
- Environmentally Preferable:
- Cdt:50
AeraSeal is a 114 µm thick, hydrophobic porous film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth.
- Minimise cross-contamination, spillage and evaporation
- Allow uniform air and CO2 exchange for all wells, unlike plate lids which favour exchange for wells near the plate edge
Dimensions: 82,6×142,9 for standard size tissue culture plates.