Charging stand for one pipette
613-4123
- Couleur:blanc
- Désignation:Portoir de charge pour 1 micropipette, prise universelle
- Seq:63
- l×p×h:380×145×115 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:Renewable energy certificate; ISO 14001
- Cdt:1
When the pipette is not in use, it should be stored in an upright position in order to avoid contamination from desks and benches.
The electronic pipettes are recommended to be stored and charged on a charging stand, whenever they are not in use. In this way, the batteries always remain charged, when the work begins. Charging Stand is for storing and charging one electronic pipette, Picus or eLINE®.