Ball valves
Ball valves, for hose int. Ø 6 - 8 mm
N° de catalogue 229-7485
Fournisseur: SEMADENI
Aller à
Product Details & Documents
Three way ball valve with PP body and PTFE tips with total length of 65 mm.
Spécifications
- Désignation:Ball valves, for hose int. Ø 6 - 8 mm