Reference standards, nitrogen standard set, CRM, VHG
Catalog # PROOVHG-NCH-SET1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Cdt:10
- Désignation:Nitrogen at 0, 1, 2, 10, 20, 50, 100, 200, 500, 1000 ng/µL in isooctane
Specifications
About this item
These calibration standards are intended for use in the determination of low level nitrogen by ASTM Method D4629.
Certifications: Traceable to NIST.