Bowie-Dick type test pack
Katalognr. 489-0039
Anbieter: PHAGOGENE
Technische Daten
- Désignation:Bowie dick test pack
- Cdt:1
Technische Daten
Über diesen Artikel
Bowie-Dick is ready-to-use test package for daily penetration test of steam for sterilisers with porous charge.
- Yellow or blue free turn indicating the good extraction of the air
- Non-toxic organic ink
Certifications: Conforms to ISO 11140-4; BSI Kitemark certified.