ContactUsLinkComponent
Recombinant Bet v 4 (from E. coli)
Recombinant Bet v 4 (from E. coli)
Supplier:  AbFrontier
Certificates
undefined
Recombinant Bet v 4 (from E. coli)
Supplier:  AbFrontier
Specifications
  • Type de protéine/peptide:
    Recombinant
  • Conditions de stockage:
    Store at −80 °C. Avoid frequent freeze and thaw.
  • Nom de la protéine/du peptide:
    Bet v 4
  • Purity:
    >95% by SDS-PAGE
  • Molecular weight:
    14 kDa
  • Formulation:
    Liquid, 20 mM HEPES, pH7,5
Specifications
Related Information
Frequently Bought Together