Cytology swabs, Histobrush®
N° de catalogue PMPC25-2199
Fournisseur: PURITAN MEDICAL
Spécifications
- Ø:5 mm
- Désignation:Cytology brushes
- Longueur:178 mm
- Cdt:1.000
Spécifications
À propos de cet article
These cytology brushes feature tapered tips made out of soft nylon bristles and a long PS handle.
Used for the collection of cervical specimens during pap smear tests, as well as buccal cells, for DNA lineage.
Certifications: ISO 9001:2008, 13485:2003, FDA GMP’s and 510K approved.
Informations de commande: A certificate of compliance is available upon request.