Human plasma, multiple sclerosis (MS)
Plasma, multiple sclerosis (MS), cryopreserved, 2 mL
N° de catalogue PB006C-1-MS
Fournisseur: Charles River Laboratories Cell Solutions
Human plasma, multiple sclerosis (MS)
N° de catalogue PB006C-1-MS
Fournisseur: Charles River Laboratories Cell Solutions
Numéro CAS:
Aller à
Product Details & Documents
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of multiple sclerosis (MS). Collected from IRB-consented donors diagnosed with MS.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Spécifications
Spécifications
- Cdt:2 mL
- Désignation:Plasma, multiple sclerosis (MS), cryopreserved